Learn More
BINDER™ ICH Light Module with LQC
ICH-Q1B light module with light-dose control
Brand:  BINDER™ 8012-2444
 View more versions of this product
Product Code. 30238201
  • 62500.47 DKK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Module consists of:
  • VIS cassette
  • UVA cassette
  • Spherical VIA sensor
  • Spherical UVA sensor
  • Independent power supply
Specifications
Specifications
KB PRO 720, KBF 720, KBF PRO 720
Glass
Lighting
LQC
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ ICH Light Module with LQC > For use with KB PRO 720, KBF 720, KBF PRO 720