Learn More
Novus Biologicals IFI44 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand:  Novus Biologicals H00010561-B01P
Product Code. 18310389
  • 4509.60 DKK / 0.05mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
IFI44 Polyclonal antibody specifically detects IFI44 in Human samples. It is validated for Western Blot
Specifications
Specifications
IFI44
Polyclonal
Western Blot 1:500
NP_006408.2
Mouse
Protein A purified
RUO
10561
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.4)
interferon-induced protein 44, interferon-induced, hepatitis C-associated microtubular aggregate protein(44kD), MTAP44p44Microtubule-associated protein 44, P44
IFI44 (NP_006408.2, 1 a.a. - 444 a.a.) full-length human protein. MAVTTRLTRLHEKILQNHFGGKRLSLLYKGSVHGFRNGVLLDRCCNQGPTLTVIYSEDHIIGAYAEESYQEGKYASIILFALQDTKISEWKLGLCTPETLFCCDVTKYNSPTNFQIDGRNRKVIMDLKTMENLGLAQNCTISIQDYEVFRCEDSLDERKIKGVIELRKSLLSALRTYEPYGSLVQQIRILLLGPIGAGKSSFFNSVRSVFQGHVTHQALVGTNTTGISEKYRTYSIRDGKDGKYLPFILCDSLGLSEKEGGLCRDDIFYILNGNIRDRYQFNPMESIKLNHHDYIDSPSLKDRIHCVAFVFDASSIQYFSSQMIVKIKRIRRELVNAGVVHVALLTHVDSMDLITKGDLIEIERCEPVRSKLEEVQRKLGFALSDISVVSNYSSEWELDPVKDVLILSALRRMLWAADDFLEDLPFEQIGNLREEIINCAQGKK
0.05 mg
Primary
Human
Purified