missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgG1 Rat Uncoated ELISA Kit

For quantitative detection of rat IgG1.

4075.00 DKK - 6750.00 DKK

Specifications

Assay Sensitivity 6.25ng/mL
Sample Type Cell Culture Supernatant, Plasma, Serum
Target Species Rat
For Use With (Application) ELISA
Storage Requirements 2°C to 8°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15530927
View Documents
Invitrogen™
88-50500-77
20 x 96 Tests
6750.00 DKK
Pack of 20
Please to purchase this item. Need a web account? Register with us today!
15550927
View Documents
Invitrogen™
88-50500-88
10 x 96 Tests
4075.00 DKK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Rat Immunoglobulin G1 (IgG1) Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of rat IgG1. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Specifications

Specifications

6.25ng/mL
Rat
2°C to 8°C
IgG1
Cell Culture Supernatant, Plasma, Serum
ELISA
Immunoglobulin G
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ IgG1 Rat Uncoated ELISA Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.