Learn More
Honeywell Safety Products™ Ignite Glasses
Lightweight, frameless, distortion free, unobstructed vision safety glasses. Honeywell Safety Products™ Ignite Safety Glasses offer a comfortable and secure fit.
56.20 DKK - 650.50 DKK
Specifications
Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1775, EC Attestation Number EC1775 |
---|---|
Product Type | Safety glasses |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Frame Material | Polyamide |
Product Code | Brand | Frame Color | Lens Tint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Frame Color | Lens Tint | Price | Quantity & Availability | ||||||
12320160
|
Honeywell Safety Products™
1017081 |
Red, Silver | N/A | N/A | N/A | ||||||
12970345
|
Honeywell Safety Products™
1017085 |
Black, Silver | N/A | N/A | N/A | ||||||
11847070
|
Honeywell Safety Products™
1017085 |
Black, Silver | N/A | N/A | N/A | ||||||
12607363
|
Honeywell Safety Products™
1017080 |
Red, Silver | N/A | N/A | N/A | ||||||
12617363
|
Honeywell Safety Products™
1017082 |
Red, Silver | Gray | N/A | N/A | N/A | |||||
12647363
|
Honeywell Safety Products™
1017087 |
Black, Silver | Gray | N/A | N/A | N/A | |||||
Description
- Nine-base, wraparound lens design.
- Close-to-the-face protection and superior coverage.
- Lightweight frameless design.
- All-day comfort and180° of distortion free, unobstructed vision.
- Adjustable nose-piece.
- Reduced slippage and personalized, comfortable, secure fit.
Specifications
EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1775, EC Attestation Number EC1775 | |
1 Ea. | |
Polyamide |
Safety glasses | |
Polycarbonate |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Honeywell Safety Products™ Ignite Glasses