Learn More
Imidazole, 0.5M buffer soln., pH 6.5, Thermo Scientific™
457.00 DKK - 702.00 DKK
Specifications
CAS 288-32-4
Concentration 0.5 M
pH 6.5
Physical Form Liquid
Molecular Formula C3H4N2
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15444619
View Documents
Thermo Scientific Alfa Aesar
J62207.AE
100 mL
457.00 DKK
100mL
15454619
View Documents
Thermo Scientific Alfa Aesar
J62207.AK
250 mL
702.00 DKK
250mL
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
288-32-4
6.5
C3H4N2
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
0.5 M
Liquid
MFCD00005183
N1C=CN=C1
68.08
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Keep cold
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Imidazole, 0.5M buffer soln., pH 6.5, Thermo Scientific™