Learn More
Thermo Scientific™ Imject™ Maleimide-Activated BSA
Prepare hapten-carrier conjugates with cysteine-containing peptides and other thiolated antigens using this sulfhydryl-reactive bovine serum albumin.
Brand:  Thermo Scientific™ 77116
 View more versions of this product
Product Code. 11874061
  • 1480.00 DKK / 2mg
Explore more special offers
Description
Show More
Specifications
Specifications
Upon receipt store product at 4°C. Product is shipped at ambient temperature.
2 mg
Imject™
Imject Maleimide Activated Bovine Serum Albumin
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberYK384995 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number77116
Lot NumberSPECS Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number77116
Lot NumberYK378011 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number77116
Lot NumberYD363148 Certificate TypeCertificate of Analysis Date29/04/2023 Catalog Number77116
Lot NumberXJ360791 Certificate TypeCertificate of Analysis Date04/11/2022 Catalog Number77116
Product Title
Thermo Scientific™ Imject™ Maleimide-Activated BSA > Activated BSA; 2mg