Learn More
Fisherbrand™ Immersion Basket with Handle
Facilitates complete immersion within S-line Ultrasonic Cleaner baths. Fisherbrand™ Immersion Baskets with Handle are manufactured from stainless steel.
Brand:  Fisherbrand™ 1026584
 View more versions of this product
Product Code. 11778728
  • 2835.00 DKK / Each
Estimated Shipment: 20-06-2025
Explore more special offers
Add to basket
Description
Description
Designed for the complete immersion of items and parts in Fisherbrand™ 112xxx Series Dual Frequency Ultrasonic Cleaners. Choose the Fisherbrand™ Stainless Steel Immersion Basket with Handles designed for the specific ultrasonic cleaner being used.
  • Mesh bottom
  • One handle on each end
  • Functional and modern design
  • Available for a variety of capacities
Specifications
Specifications
Basket
Ultrasonic Bath, Elmasonic Xtra Basic 550 55L
Stainless Steel
Videos
  • Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Fisherbrand - Ultrasound for laboratory applications and for intensive cleaning
    Video Player is loading.
    Current Time 0:00
    Duration 4:02
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 4:02
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Fisherbrand Ultrasound for laboratory applications and for intensive cleaning
Product Title
Fisherbrand™ Immersion Basket with Handle > For X-tra Basic 550 Ultrasonic Baths