Learn More
SI Analytics™ Immersion Thermostat and Thermostatic Bath
SI Analytics™ Immersion Thermostat and Thermostatic Bath is supplied in a stainless steel container with one manual gauge slide.
Brand:  SI Analytics™ 285418547
57473.71 DKK valid until 2025-07-31
Use promo code "24075" to get your promotional price.
Product Code. 11904729
  • 67310.00 DKK / Each
The Ultimate Lab Appliances Promotion
Use promo code 24075 at checkout
View Available Promotions
Only null left
In Stock
Description
  • Includes an immersion thermostat and thermostatic bath (stainless steel container with one manual gauge slide), basic configuration for the attachment of one flow-through cooler
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.
    Promo Code: 24075
    Expires2025-07-31
    Days
    15
    Hours
    17
    Minutes
    29
    Seconds
    46
Product Content Correction
Product Title
SI Analytics™ Immersion Thermostat and Thermostatic Bath > Voltage: 230 V