missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Tubes and Stoppers

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.

Brand:  Thermo Scientific™ 348801

 View more versions of this product

Product Code. 11351785

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Polyethylene
12 mm
3000
12 mm Nunc-Immuno Tubes
Stopper
500
Not Applicable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number165453 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number348801
Lot Number182017 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number348801
Lot Number182905 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number348801
Lot Number181580 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number348801
Lot Number169850 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number348801
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Tubes and Stoppers > Nunc-Immuno, 12

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.