missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ImmunoCult™-ACF Dendritic Cell Medium
Medium for the culture and differentiation of human monocytes into dendritic cells and their maturation
1614.00 DKK - 5676.00 DKK
Specifications
Cell Type | Dendritic Cells, Monocytes |
---|---|
For Use With (Application) | Cell Culture; Differentiation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17137651
|
STEMCELL Technologies
10987 |
100 mL |
1614.00 DKK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16655635
|
STEMCELL Technologies
10986 |
500 mL |
5676.00 DKK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ImmunoCult™-ACF Dendritic Cell Medium and Supplements, sold individually or as a kit, have been developed for the in vitro culture and differentiation of human monocytes into dendritic cells (DCs)
- ImmunoCult™-ACF Dendritic Cell Medium is a serum-free and animal component-free medium optimized for the in vitro culture and differentiation of human monocytes into DCs
- ImmunoCult™-ACF Dendritic Cell Differentiation Supplement contains a combination of animal component-free recombinant human cytokines formulated to support the differentiation of immature DCs from human monocytes
- The ImmunoCult™ Dendritic Cell Maturation Supplement has been formulated to support maturation of immature DCs to mature DCs
Specifications
Dendritic Cells, Monocytes | |
Cell Culture; Differentiation |