missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IMPDH2 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67663-1-IG-150UL
This item is not returnable.
View return policy
Description
The IMPDH2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IMPDH2. This antibody recognizes human, mouse, rat antigen. The IMPDH2 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
IMPDH2 | |
Monoclonal | |
500 μg/mL | |
IMPD2; IMPDH-II | |
Recombinant Protein | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
56 kDa | |
Liquid |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
1B1G8 | |
Unconjugated | |
Mouse | |
Protein A purification | |
RUO | |
3615 | |
Store at -20°C. Stable for one year after shipment. | |
Monoclonal Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IMPDH2 Mouse Monoclonal Antibody, Proteintech > Unconjugated; 150 μL
Spot an opportunity for improvement?Share a Content Correction