missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Increased Protection Gloves

Product Code. 15826791
Click to view available options
Size 11
This item is not returnable. View return policy
This item is not returnable. View return policy

Euro Protection™ Increased Protection Gloves are water-repellent and offer an excellent grip in cold, moist environments.

  • Water-repellent
  • Flexible
  • Increased dexterity
  • Excellent grip
  • Suitable for light duty in cold and moist environments, handling of abrasive parts and slippery objects, assembly and mechanical work
  • Nylon-based seamless knit
  • PVC-HPT coating
  • Terry cotton liner
  • Elastic wrist

Specifications

Certifications/Compliance (Europe) EN388:20033232EN511:2006020EN420EN388:2016in process)
Description Nylon-based seamless knit glove
Coating Material PVC-HPT
Color Gray
Material Nylon
Size 11
For Use With (Application) Light duty in cold and moist environments, handling of abrasive parts and slippery objects, assembly and mechanical work
Liner Material Terry cotton
Quantity 10/Pk
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'