Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Brand:  Fisherbrand™ FB67763
 View more versions of this product
Product Code. 15525049
  • 2355.00 DKK / 1 unit
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
Specifications
Specifications
E2
500 g
Cylindrical
Stainless Steel
500 g
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class E2 > 500g