Learn More
Eppendorf™ Innova 40R Benchtop Shaker Bundle
The Eppendorf Innova 40 and 40R (refrigerated) benchtop incubator shakers can take flasks up to 3 L in a compact footprint. The user interface allows for easy programming of speed, temperature, and time, and samples can be observed at all times through the self-supporting transparent lid.
Brand: Eppendorf™ M1299-0296
102028.10 DKK valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
mAb-Discovery Bundle: Molecular Biology Benchtop Solution, for plasmid construction and expression in bacteria (small volume). Bundle consists of: 1x New Brunswick™ Innova™ 40R (refrigerated shaker, 25 mm/1 in orbit), 1x universal platform, 1x tube rack small, each 3x clamps for 250 mL/500 mL/1 L Erlenmeyer flasks, 230 V/50 Hz (EU)
- Triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble-free operation for years
- Compact benchtop model – fits easily on the bench and holds flasks up to 3 L
- Clear cover allows easy viewing of the cultures and lifts easily to access all samples
- Certified. Meets cETL, UL, CSA and CE standards
Specifications
25 mm | |
230 V | |
50 Hz | |
0.01 to 99:59 hr., Continuous |
25 to 500 rpm | |
Ambient -30° to 110°C | |
76.2 x 55.9 x 61 cm (22 x 30 x 24 in.) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.