Learn More
New Brunswick Scientific™ Innova™ 42 Incubator Shakers
Use these versatile incubator shakers for a variety of applications. New Brunswick Scientific™ Innova™ 42 Incubator Shakers provide exceptional convenience allowing you to simultaneously shake and static incubate cultures under the same conditions.
Brand:  New Brunswick Scientific™ M1335-0012
85449.52 DKK valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 11350025
  • 109742.22 DKK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Stackable or benchtop incubator shaker with optional refrigeration
  • Use on or under the bench, or double stacked to save space
  • Optional culture drawer, gassing manifold, remote alarm, and humidity monitor are available
  • Cast iron triple-eccentric counterbalanced drive provides years of dependable operation
  • Programmable Innova controls automatically changes temperature, speed and optional photosynthetic and UV germicidal lights at timed intervals
  • Large viewing window and internal light provide easy viewing of samples without the need to open the door
  • Built-in water reservoir humidifies chamber to reduce sample evaporation, while also protecting unit from spills and including a drain for easy cleaning
  • Versatile accessory platforms (sold separately) accommodates flasks up to 4 L as well as test tube racks and microplate holders
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mm
230 V, 50 Hz
4 L
50 Hz
131 cm
Innova 42
25 to 400 rpm
230 V
Incubator Shaker
75.4 cm
62 cm
0.01 to 99:59 hr.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products