missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Innova 42R Microbiology Shaker Bundle

For total versatility, the Eppendorf Innova 42R (refrigerated) incubator shaker is stackable, can be placed in numerous locations, and have a number of user options. These include: a static culture drawer, gassing manifold, remote alarm, and humidity monitor and drip pan.

Brand:  Eppendorf™ M1335-0216

109286.50 DKK valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 17159114

  • 140314.02 DKK / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

mAb-Discovery Bundle: Molecular Biology Versatility Solution, for plasmid construction and expression in bacteria (medium volume). Bundle consists of 1x New Brunswick™ Innova™ 42R (refrigerated shaker, 25 mm/1 in orbit), 1x universal platform, 1x tube rack medium and each 3x clamps for 250 mL/500 mL/1 L Erlenmeyer flasks, 230 V/50 Hz (EU)

  • High capacity in a small footprint: for example, 4x 5 L or 2x 6 L flasks on universal platform; device footprint 4731 cm2 /733 in2 (63.5 cm/25 in. width, 74.5 cm/29.3 in. depth)
  • Ergonomical and safe interface handling: Top mounted controls and display make bending down unnecessary during handling, avoids accidental parameter changes, and can be reached easily even in a double stack
  • Easy to clean: Spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation); airflow UV-germicidal light optional
  • Lab position flexibility: Lab position on or under bench, 2x stackable
  • Parallel static cultivation: One height adjustable static shelf included, optional culture drawer and additional shelves available
  • Visual sample control: Large viewing window and internal light provide easy viewing of samples without the need to open the door
  • Reliable 24/7 with high loads: Proven and tested Innova triple eccentric counterbalanced drive (three weight-supporting shafts), max. load 15 kg
  • Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
  • Options and Accessories: Orbit 19 or 25 mm, photosynthetic light bank, UV germicidal light, culture drawer for evaporation protection of Agar plates, additional static shelves (one included), humidity monitoring, remote monitoring with VisioNize™ Lab Suite and more
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mm
230 V
74.5 x 63.5 x 81.8 cm (25 x 29.3 x 32.8 in.)
25 to 400 rpm (25 to 300 rpm when using flasks >500 mL in stacked devices)
50 Hz
0.1 to 99.9 hr., Continuous
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers