Learn More
NORDIC SERVICE GROUP™ Installation IQ/OQ on Memmert Chambers
Installation and Operational Qualification (IQ/OQ) on Memmert Chambers supported by Nordic Service Group.
Brand: NORDIC SERVICE GROUP™ MEMDKIQOQ3
Description
IQ (Installation Qualification):
This qualification ensures the -test object- appliance is inspected for completeness of delivery and operational readiness. It also documents all essential specifications and parameters stipulated by the manufacturer, verifies compliance with safety regulations, and identifies the individuals instructed in the basic functions and operation of the appliance.
OQ (Operational Qualification):
This qualification verifies and tests the operational and production readiness of the -test object- appliance.
On-site testing includes:
- All quality or process-relevant alarm and switching functions.
- All quality or process-relevant control functions.
- Temperature distribution and measurement of humidity, CO2, O2, pressure, and illuminance (depending on the device type and equipment).
The necessary setpoint settings (parameters) for the test must be configured as described in the respective operating manuals.
Note: The instrument must be in its designated place. Moving work beyond what one person can do will incur additional costs.
Specifications
Memmert Chambers | |
IQ/OQ Validation Service |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.