Learn More
Memmert™ Interior IP68 Socket for UF260
Interior IP68 socket that can be turned on and off with a switch.
Brand: Memmert™ UF260R3
This item is not returnable.
View return policy
Description
- Can only be ordered with limited temperature range - max. +60°C)
- Electrical Requirements: 230 V, 2 A, 500 W
- Can be switched off with the On/Off switch
- Cannot be switched individually
- Moisture tight IP68
- Requires option A8 against surcharge
Specifications
UF260 Oven | |
Interior Socket | |
230 V | |
2 A |
Product Content Correction
Product Title
Memmert™ Interior IP68 Socket for UF260 > For Use With: UF260
Spot an opportunity for improvement?Share a Content Correction