missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1485.92 DKK - 3498.83 DKK
Specifications
Antigen | INTS6 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310477
|
Bio-Techne
NBP3-04624-20UL |
20 μL |
1485.92 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18391137
|
Bio-Techne
NBP3-04624-100UL |
100 μL |
3498.83 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INTS6 Polyclonal specifically detects INTS6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
INTS6 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DBI1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 26, DICE1DBI-1DDX26ADDX26DEAD box protein, DKFZp434B105, HDB, INT6, integrator complex subunit 6, Notchl2, Protein DDX26, Protein deleted in cancer 1, RNA helicase HDB | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human INTS6 (NP_036273.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Growth and Development, Neuronal Cell Markers, Tumor Suppressors | |
PBS with 50% glycerol, pH7.3. | |
26512 | |
IgG | |
Affinity purified |