Learn More
Invitrogen™ Yeast Deletion Heterozygous Essential, Complete
Click to view available options
Quantity:
16 Plates
Unit Size:
Each
Description
Each yeast deletion clone is supplied as a stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418).
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
Product Type | Yeast Deletion Heterozygous Essential, Complete |
Libraries | Yeast deletion library created by a collaboration of 8 North American and 8 European laboratories composing the Saccharomyces Genome Deletion Project |
Culture Type | Stab culture in yeast peptone dextrose (YPD) agar containing Geneticin™ (G418) |
Format | Complete Set |
Quantity | 16 Plates |
Product Content Correction
Product Title
Invitrogen™ Yeast Deletion Heterozygous Essential, Complete >
Spot an opportunity for improvement?Share a Content Correction