Learn More
Mettler Toledo™ Iodide Half Cell Electrode
Brand: Mettler Toledo™ 51107680
9251.59 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Specifications
Solid State Membrane | |
0°C to 80°C | |
1 to 13 |