missing translation for 'onlineSavingsMsg'
Learn More
Iodine, 0.05M/l Solution, Scharlab Product Code.: 11394184

Iodine, 0.05M/l Solution, Scharlab

Product Code. 11394184
1 L
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy

Product Code. 11394184

Brand: Scharlab YO00231000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in analytical chemistry as a titrant in volumetric analysis.

This volumetric solution was checked by means of potentiometric methods using a sodium thiosulfate standard solution, that was also checked against Scharlau's potassium iodate volumetric standard. Scharlau's volumetric standards are directly traceable to the Standard Reference Materials from NIST (National Institute of Standards and Technology, USA).

  • Molecular Formula: I2
  • Molar Mass: 253.81g/mol
  • CAS: 7553-56-2
  • EIN ECS No.: 231-442-4
  • Factor: 0.995–1.005

Specifications

CAS 7553-56-2
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
Composition 1mL = 0.0127g I2
Density 1.02g/cm3
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Formula Weight 253.81g/mol
Traceability to NIST Traceable to NIST
Quantity 1 L
Physical Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine, 0.05M/l Solution, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.