missing translation for 'onlineSavingsMsg'
Learn More

Iodophenpropit dihydrobromide, Tocris Bioscience™

Product Code. 18043902
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective H3 antagonist

Selective H3 antagonist with high affinity (KD = 0.3 nM).

Chemical Identifiers

CAS 145196-87-8
Molecular Formula C15H21Br2IN4S
Molecular Weight (g/mol) 576.133
InChI Key BOSOGNBLIWPCMS-UHFFFAOYSA-N
Synonym iodophenpropit dihydrobromide, iodophenpropit hydrobromide, 3-1h-imidazol-4-yl propyl n-2-4-iodophenyl ethyl carbamimidothioate dihydrobromide, n-2-4-iodophenyl ethyl-s-3-4 5-imidazolyl propyl isothiourea dihydrobromide
PubChem CID 24978528
IUPAC Name 3-(1H-imidazol-5-yl)propyl N'-[2-(4-iodophenyl)ethyl]carbamimidothioate;dihydrobromide
SMILES C1=CC(=CC=C1CCN=C(N)SCCCC2=CN=CN2)I.Br.Br

Spécification

Quantity 10 mg
Formula Weight 576.13
Percent Purity >98%
Chemical Name or Material Iodophenpropit dihydrobromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'