Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2, Humidity and Temperature
For one free-selectable CO2, humidity and temperature value
31940.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17139994
|
Memmert™
ICO240D38897 |
ICO240 |
31940.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17160025
|
Memmert™
ICO50D38897 |
ICO50 |
31940.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17169924
|
Memmert™
ICO105D38897 |
ICO105 |
31940.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199954
|
Memmert™
ICO150D38897 |
ICO150 |
31940.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Includes temperature distribution survey at Memmert
- For 27 measuring points to DIN 12880:2007-05
- PQ check list as support for validation by customer.
Specifications
IQ/OQ Document with Device-specific Works Test Data |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data - CO2, Humidity and Temperature