Learn More
Memmert™ IQ/OQ Document with Test Data for SF260
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
Brand: Memmert™ SF260D00127
Description
- Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
- PQ check list as support for validation by customer
Specifications
SF260 Sterilizer | |
IQ/OQ Documentation | |
Test Data |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.