Learn More
Memmert™ IQ/OQ Document with Test Data for SF260
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
Brand:  Memmert™ SF260D00127
Product Code. 17588175
  • 16185.00 DKK / Each
Explore more special offers
Description
Description
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
Specifications
SF260 Sterilizer
IQ/OQ Documentation
Test Data
Product Title
Memmert™ IQ/OQ Document with Test Data for SF260 > For Use With: SF260