Learn More
Memmert™ IQ/OQ Document with Test Data for SF75
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
Brand:  Memmert™ SF75D00127
Product Code. 17568395
  • 16185.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
Specifications
SF75 Sterilizer
IQ/OQ Documentation
Test Data
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Test Data for SF75 > For Use With: SF75