Learn More
Memmert™ IQ/OQ Document with Test Data for UF1060
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
Brand: Memmert™ UF1060D00127
This item is not returnable.
View return policy
Description
- Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
- PQ check list as support for validation by customer
Specifications
UF1060 Oven | |
IQ/OQ Documentation | |
Test Data |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Test Data for UF1060 > For Use With: UF1060
Spot an opportunity for improvement?Share a Content Correction