Learn More
Memmert™ IQ/OQ Document with Test Data for UF55
IQ/OQ document with temperature distribution survey for one free-selectable temperature value.
Brand:  Memmert™ UF55D00127
Product Code. 17559445
  • 16185.00 DKK / Each
This item is not returnable. View return policy
Description
Description
  • Includes temperature distribution survey with 9 temperature measuring points (size 30) and 27 measuring points (sizes 55 - 1060) to DIN 12880:2007-05 (further temperature values on demand)
  • PQ check list as support for validation by customer
Specifications
Specifications
UF55 Oven
IQ/OQ Documentation
Test Data
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Test Data for UF55 > For Use With: UF55