Learn More
Palintest™ Iron HR Test Kits
Robust and reliable iron HR test kit for photometers.
Brand:  Palintest™ PM156
Product Code. 11923617
  • 312.00 DKK / Pack of 50
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Palintest Iron Test Reagent. 0 to 10 ppm, 50 Tests/Kit
  • The Palintest Iron HR test is based on a single tablet reagent containing an alkaline thioglycollate
  • The test is carried out simply by adding a tablet to a sample of the water under test
    • Iron (HR), Photometer Reagents, 0 - 10 mg/L Fe
Specifications
Specifications
50
0 to 10mg/L
Iron HR Test Kit
50 Pack
50 Tests
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Palintest™ Iron HR Test Kits > Range: 0 to 10mg/L