missing translation for 'onlineSavingsMsg'
Learn More
Iron solution for ICP 10g/L HCl matrix, Fisher chemical™ Product Code.: 12976294
compliance-icons

Iron solution for ICP 10g/L HCl matrix, Fisher chemical™
BIOPHARMA

Product Code. 12976294 Shop All Fisher Chemical Products
100 mL, Bottle
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy

Product Code. 12976294

Brand: Fisher Chemical S 219 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
Analyte or Component Names Fe
Concentration 10±0.02 g/L
Type Mass Spectrometry Standard
Element Fe
Solvent or Matrix HCl
Quantity 100 mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1103661 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog NumberS 219 100
Lot Number1010628 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberS 219 100
Lot Number1059283 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberS 219 100
compliance-icons
Product Identifier
  • Single-element Standard Solution for ICP. Iron (Fe) 10000mg/l in HCl 5%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron solution for ICP 10g/L HCl matrix, Fisher chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.