Learn More
R&D Systems™ Irradiated Mouse Embryonic Fibroblasts (5 vials)
For use as a feeder layer when culturing embryonic or induced pluripotent stem cells
Brand: R&D Systems™ PSC001
2844.00 DKK valid until 2025-07-30
Use promo code "24078" to get your promotional price.
Description
Mitotically inactive mouse embryonic fibroblasts (MEFs) support the growth and maintenance of stem cell pluripotency by secreting necessary soluble factors and by providing a substrate for stem cell attachment.
- Ready to use
- MEFs support undifferentiated growth of embryonic and induced pluripotent stem cells
- Inactivated by gamma irradiation to avoid chemical contamination
- Consistent feeder layer to reduce experimental variation
- Free of microbial and mycoplasma contamination
Specifications
6 x 10e6 Cells/Vial, 5 Vials |
Promotions
-
R&D Systems
,
Tocris
Ultimate Promo May
Empowering Research: The Right Reagents for Every StepPromo Code: 24078Expires: 2025-07-30