Learn More
WTW™ IS6 Stirrer for WTW™ OxiTop™ System
Brand: WTW™ 208140
7162.52 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
IS6 Stirrer | |
For 6 stirring | |
6 Measuring units | |
Universal power supply |
Product Content Correction
Product Title
WTW™ IS6 Stirrer for WTW™ OxiTop™ System > Voltage: 100 to 240VAC; 6 Measuring units
Spot an opportunity for improvement?Share a Content Correction