missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ISO Horizontal Safety Sign - Warning: Hot Surface
Safety sign for hazard identification.
Brand: Brady™ 828908
145.35 DKK valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sign compliant to ISO 7010
- Improves workplace safety
Specifications
STEN W017-150x50-AL-CRD/1 | |
Rectangular | |
50 x 150 mm | |
Black on Yellow |
Aluminum | |
English | |
Warning Hot Surface |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction