Learn More
Labconco™ Isolation Valve
Shorten the time necessary to reestablish working vacuum levels and to provide a means for determining end point. Labconco™ Isolation Valve is for use with FreeZone™ Stoppering Tray Dryers and FreeZone™ Bulk Tray Dryers.
Brand:  Labconco™ 7761500
Product Code. 15183085
  • 31850.00 DKK / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Isolates the Tray Dryer from the Freeze Dry System to shorten the time necessary to reestablish working vacuum levels and to provide a means for determining end point
  • Consists of a valve, valve handle, clamps, couplings and installation instructions
  • Mounts underneath the support stand included with the Tray Dryer
  • May be installed before or after the Tray Dryer is installed on the Freeze Dry System
  • For use with Stoppering Tray Dryers 7806020, 7806040 and 7806030 and Bulk Tray Dryers 7806060 and 7806070.
  • The Isolation Valve and 6-Port Tray Dryer Manifold 7726500 may not be field installed together.
Specifications
Specifications
Isolation Valve
8.65 in.
2.3 kg
7.75 in.
5 lb.