missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl methanesulfonate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Density | 1.1450g/mL |
Boiling Point | 82.0°C (6.0 mmHg) |
Flash Point | 104°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass Bottle |
Linear Formula | CH3SO3CH(CH3)2 |
Refractive Index | 1.4180 to 1.4210 |
Quantity | 100 g |
Show More |
Safety and Handling
Product Identifier
- Isopropyl methanesulfonate
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'