Learn More
KGW Isotherm™ Isotherm Borosilicate Glass Insulating Lid
Brand:  KGW Isotherm™ 17211
 View more versions of this product
Product Code. 10566232
  • 293.00 DKK / Each
Only null left
Estimated Shipment: 30-07-2025
Specifications
1000 mL
Flexible Applications In Laboratory
1
Borosilicate Glass
Dewar Container; Type 9 Plugs
Insulating Lid
Certificates
Promotions
Promotions
Product Content Correction
Product Title
KGW Isotherm™ Isotherm Borosilicate Glass Insulating Lid > Capacity: 1000mL; For use with: Type 9 Plugs