missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IUPAC pH Standard pH 4.005, Hach Lange™
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- Traceable to IUPAC pH scale and NIST
- Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
- The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference material
- Shelf life of 48 months from production date
Specifications
Specifications
pH | 4.005 |
Physical Form | Liquid |
Quantity | 500 mL |
Traceability to NIST | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IUPAC pH Standard pH 4.005, Hach Lange™ >
Spot an opportunity for improvement?Share a Content Correction