missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.
Solubility
Soluble in methyl ethyl ketone (50 mg/ml), and DMSO (50 mM).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
It is used as a positive allosteric modulator of the α7 neuronal nicotinic acetylcholine receptor.
Solubility
Soluble in methyl ethyl ketone (50 mg/ml), and DMSO (50 mM).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.

Specifications
Specifications
Quantity | 250mg |
UN Number | UN2811 |
Merck Index | 14,5248 |
Solubility Information | Soluble in methyl ethyl ketone (50mg/ml),and DMSO (50 mM). |
Formula Weight | 875.11 |
Chemical Name or Material | Ivermectin |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X04I055 | Certificate of Analysis | 13/10/2022 | J62777.03,J62777.MD |
F17Z017 | Certificate of Analysis | 08/12/2021 | J62777.03,J62777.MD |
Y02B007 | Certificate of Analysis | 07/12/2021 | J62777.03,J62777.MD |
F14X010 | Certificate of Analysis | 07/12/2021 | J62777.03,J62777.MD |
P16E062 | Certificate of Analysis | 07/12/2021 | J62777.03,J62777.MD |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ivermectin, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction