Learn More
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap is specially designed for use with vacuum manifolds and Schlenk lines.
Brand: GPE Scientific™ JCF/250
This item is not returnable.
View return policy
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap
Specifications
250 mL | |
Vacuum manifolds and Schlenk lines | |
Fine thread tap | |
Sample Flask |
Product Content Correction
Product Title
GPE Scientific™ J Young Sample Flask, Cylindrical with Fine Thread Tap > Capacity: 250mL
Spot an opportunity for improvement?Share a Content Correction