Learn More
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
Brand:  GPE Scientific™ CSY/100
 View more versions of this product
Product Code. 15834744
  • 1145.00 DKK / Each
Only null left
Estimated Shipment: 15-10-2025
Description
  • Designed with a side arm for evacuating or filling with gases
  • For connection to Schlenk lines
  • Includes O-ring tap
  • Includes cold finger plain arm
Specifications
Specifications
100 mL
O-ring tap and cold finger plain arm
Sample Flask
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GPE Scientific™ J Young Sample Flask with O-Ring Tap and Cold Finger Plain Arm > Capacity: 100mL