Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm is designed for use with Schlenk lines and a vacuum manifold and available in various capacities.
959.00 DKK - 1060.00 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15834734
|
GPE Scientific™
CSB/25 |
25 mL |
974.00 DKK
Each |
Estimated Shipment: 01-09-2025
Log in to see stock. |
||||||
15854734
|
GPE Scientific™
CSB/50 |
50 mL |
1005.00 DKK
Each |
Estimated Shipment: 14-08-2025
Log in to see stock. |
||||||
15814734
|
GPE Scientific™
CSB/100 |
100 mL |
1015.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||||
15844734
|
GPE Scientific™
CSB/250 |
250 mL |
1045.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||||
15864734
|
GPE Scientific™
CSB/500 |
500 mL |
959.00 DKK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||||
15824734
|
GPE Scientific™
CSB/1000 |
1000 mL |
1060.00 DKK
Each |
|
||||||
Description
- Designed with a side arm for evacuating or filling with gases
- For connection to Schlenk lines
- Round-bottomed shape
- Includes plain arm
- Includes O-ring tap
Specifications
O-Ring Tap and Plain Arm | |
Vacuum/Schlenk Flask |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm