Learn More
Lenz Laborglasintrumente Jacketed Extractor Heads Acc. To Knöfler-Böhm
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1715.00 DKK - 3220.00 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10315741
|
Lenz Laborglasintrumente
05410125 |
30 mL |
1715.00 DKK
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10768271
|
Lenz Laborglasintrumente
05410158 |
500 mL |
3220.00 DKK
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
10819510
|
Lenz Laborglasintrumente
|
250 mL |
2385.00 DKK
Each |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
Description
- Jacketed extractors acc. to DIN 12604
- Made of DURAN™ tubing
Specifications
Duran Glass |