Learn More
Justrite™ Brass Faucet
Self-closing safety faucets prevents flashback of fire into drum
Brand: Justrite™ 08540
443.18 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
- Features polytetrafluoroethylene (PTFE) seal
- Adjustable models screw tightly into drum
- Faucet extensions provide extra dispensing accuracy and prevents spills
Certifications
FM Global tested and approved
Specifications
FM | |
Self Close | |
0.75 in. |
Brass Control Flow Lab Safety Faucet | |
Brass | |
1.90 cm |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now