Learn More
Nordvlies Katrin Inclusive Folded Toilet Tissue Dispenser
Katrin Inclusive Folded Toilet Tissue Dispenser is well-suited for areas with frequent use.
442.00 DKK
Specifications
Product Type | Toilet Paper Dispenser |
---|---|
Dimensions (L x W x H) | 307 x 158 x 133 mm |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
15711097
|
Nordvlies
92582 |
White |
442.00 DKK
Each |
Estimated Shipment: 18-11-2025
Log in to see stock. |
||||||
15721097
|
Nordvlies
92605 |
Black |
442.00 DKK
Each |
Estimated Shipment: 18-11-2025
Log in to see stock. |
||||||
Description
- Folded toilet tissue dispenser
- Hygienic - feeds out one sheet at a time (reduces cost in use)
- Inspection window shows when it is time to refill the paper
- Simple to install, load and maintain
- Made from ABS Plastic
- High capacity over 600 folded toilet paper sheets
- Lockable
Specifications
Toilet Paper Dispenser | |
307 x 158 x 133 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Nordvlies Katrin Inclusive Folded Toilet Tissue Dispenser