missing translation for 'onlineSavingsMsg'
Learn More

KBTBD7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-05059-100UL

 View more versions of this product

Product Code. 18313225

  • 3498.83 DKK / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

KBTBD7 Polyclonal specifically detects KBTBD7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

KBTBD7
Polyclonal
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
84078
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
DKFZP434E2318, kelch repeat and BTB (POZ) domain containing 7, kelch repeat and BTB domain-containing protein 7
Recombinant fusion protein containing a sequence corresponding to amino acids 475-684 of human KBTBD7 (NP_115514.2).
100 μL
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers