Learn More
Test and Calibration Standard 1413 μS/cm at 25 °C, WTW™
Click to view available options
Quantity:
50 mL
Unit Size:
Pack of 6
Description
- Test and calibration standard 1413 μS/cm at 25 °C
- Test resistors and calibration solutions to test the meters with conductivity function
- Calibrate the system from the device and conductivity measuring cell.
- Optimal measuring safety by PTB/NIST traceable standard solutions
- Easy device check by highly precise test resistors
- Pharmacopeia conform test kits
Specifications
Specifications
Traceability to NIST | Yes |
Solution Contains | KCL |
Physical Form | Liquid |
Concentration | 0.01 mol/L |
Color | Colorless |
Quantity | 50 mL |
Product Content Correction
Product Title
Test and Calibration Standard 1413 μS/cm at 25 °C, WTW™ >
Spot an opportunity for improvement?Share a Content Correction