Learn More
DWK Life Sciences™ KECK™ Clip for Conical Joints, Ni, KCM
KECK™ conical ground joint clamps for high temperature applications.
Brand: DWK Life Sciences™ 286337405
51.75 DKK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
For applications at very high temperatures, the KECK™ nickel-plated spring steel ground joint clamps an important addition to KECK™ conical ground joint clips made of POM (polyoxymethylene). Compared to conventional wire clamps, they are easy and safe to attach and are inherently stable.
- Easy and safe to put on and are dimensionally stable
- Good dimensional stability compared to conventional wire clamps
Specifications
100 | |
For Use at Very High Temperatures | |
NS 19 |
Nickel | |
For use at very high temperatures; Securing conical ground joints |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now