Learn More
DWK Life Sciences™ KECK™ Clip, for conical joints, from POM
The KECK™ clamp is used to secure a standard ground joint connection.
Brand: DWK Life Sciences™ 1091140
304.58 DKK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
- For conical joints
- Dimensionally stable with particularly good spring properties
- Easy, safe and quick to set up
- Increased safety in the laboratory as the glass surface is not scratched
- Marking of the individual sizes for better differentiation
Specifications
10 | |
Secure Glass-to-Glass Joints | |
NS 24 |
POM | |
Secure glass-to-glass joints | |
Green |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now