Learn More
DWK Life Sciences KECK™ O-ring
Spare part for the KECK™ adapter KA from glass olive to threaded plastic nozzle.
Brand:  DWK Life Sciences 286342108
 View more versions of this product
Product Code. 10419542
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The O-ring is a component and therefore a spare part for the KECK™ Adapter KA from glass olive to plastic olive.
  • The O-ring is a component/spare part for the KECK™ Adapter KA and is fitted between the screw cap and clamping ring for Adapter KA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sealing
100/Pk.
3 mm H
VITON
6 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
DWK Life Sciences KECK™ O-ring > VITON, diameter 6 mm