Learn More
DWK Life Sciences KECK™ O-ring
Spare part for the KECK™ adapter KA from glass olive to threaded plastic nozzle.
Brand: DWK Life Sciences 286342202
This item is not returnable.
View return policy
Description
The O-ring is a component and therefore a spare part for the KECK™ Adapter KA from glass olive to plastic olive.
- The O-ring is a component/spare part for the KECK™ Adapter KA and is fitted between the screw cap and clamping ring for Adapter KA.
Specifications
Sealing | |
100/Pk. | |
2.4 mm H |
EPDM | |
7.3 mm |
Product Content Correction
Product Title
DWK Life Sciences KECK™ O-ring > EPDM, diameter 7.3 mm
Spot an opportunity for improvement?Share a Content Correction